Would you like to log in again?
CHO-K1/CD155 STABLE CELL LINE 2VIALS
Supplier: Genscript
CD155, commonly known as PVR (poliovirus receptor) and Necl-5 (nectin-like molecule-5), is a type I transmembrane single-span glycoprotein, and belongs to the nectins and nectin-like (Necl) subfamily. CD155 was originally identified based on its ability to mediate the cell attachment and entry of po
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CHO-K1/CD155 STABLE CELL LINE 2VIALS 76510-544 M00591 Each Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty