Would you like to log in again?
CD16/CHO-K1 158V STABLE
Supplier: Genscript
CD16 is a cluster of differentiation molecule found on the surface of natural killer cells, monocytes and macrophages. It can be used to isolate populations of these cells by antibodies directed towards CD16, using fluorescent-activated cell sorting or magnetic-activated cell sorting. CD16 has been
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CD16/CHO-K1 158V STABLE 76510-556 M00597 Each Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty