Would you like to log in again?
CHO-K1/ICOS STABLE CELL L
Supplier: Genscript
protein encoded by this gene belongs to CD28 and CTLA-4 cell-surface receptor family. ICOS (Inducible T-cell COStimulator) is expressed on activated T cells. It forms homodimers and plays an important role in cell-cell signaling, immune responses, and regulation of cell proliferation. It is
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CHO-K1/ICOS STABLE CELL L 76510-568 M00649 Each Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty