Would you like to log in again?
B-AMYLOID (25-35) 5MG
Supplier: Genscript
Beta-amyloid protein (Abeta), a major component of senile plaques of Alzheimer's disease (AD) in the brain, causes elevation of the intracellular free Ca2+ level and the production of robust free radicals. Beta-amyloid 25-35 induced apoptosis, characterized by decreased cell viability, n, Size: 5mg
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
B-AMYLOID (25-35) 5MG 76510-690 RP10008-5 Each (5mg) Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty