Would you like to log in again?
CORTISTATIN 14 500UG
Supplier: Genscript
Cortistatin 14 is a somatastatin-like neuropeptide with neuronal depressant & sleep modulating properties. Cortistatin 14 is present primarily in cortex & hippocampus. In hippocampus, cortistatin 14 inhibits pyramidal cell firing & co-exists with GABA. To determine if cortistatin-14, Size: 500uG
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CORTISTATIN 14 500UG 76510-816 RP10972-0.5 Each (500µG) Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty