Would you like to log in again?
HEXA-HIS AMINO ACID SEQUENCE 5MG
Supplier: Genscript
amino acid sequence consisting of 6 His residues in a row, called His-Tag, would act as a metal binding site for a recombinant protein. His-Tag sequence can be placed on N- or -terminal of a target protein by using vectors from various commercial molecular biology companies. This Tag i, Size: 5mg
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
HEXA-HIS AMINO ACID SEQUENCE 5MG 76510-914 RP11737-5 Each (5mg) Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty