Would you like to log in again?
LH-RH (HUMAN) HYPOTHALAMIC PEPTIDE 5MG
Supplier: Genscript
LH-RH (human), a hypothalamic peptide, is involved in the regulation of reproductive functions by controlling the synthesis and release of luteinizing hormone (LH) and follicle-stimulating hormone (FSH) from the anterior pituitary gland, Size: 5mg
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
LH-RH (HUMAN) HYPOTHALAMIC PEPTIDE 5MG 76510-932 RP11937 Each (5mg) Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty