Would you like to log in again?
CHEMERIN-9 (149-157) 1MG
Supplier: Genscript
Chemerin-9 (149-157), nonapeptide 149,YFPGQFAFS157 (chemerin-9), corresponding to C terminus of processed chemerin, retained most of activity of full-size protein, with regard to agonism toward chemerinR. Natural ligand of ChemR23 (chemerinR).
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CHEMERIN-9 (149-157) 1MG 76511-078 RP20248 Each (1mg) Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty