Would you like to log in again?
HCOV-OC43 SPIKE GLYCOPROT
Supplier: Genscript
Hcov-Oc43 Spike Glycoprot, This pool is delivered in two subpools of two subpools of 168 peptides derived from a peptide scan (15mers with 11 aa overlap) through the entire Spike glycoprotein (Protein ID: P36334) of Human coronavirus OC43 (HCoV-OC43) for T cell assays.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
HCOV-OC43 SPIKE GLYCOPROT 76511-248 RP30011 Each Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty