Would you like to log in again?
SARS-COV-2 ORF8 1VIAL(25UG/PEPTIDE)
Supplier: Genscript
Sars-Cov-2 Orf8, This pool includes 28 peptides derived from a peptide scan (15mers with 11 aa overlap) through the entire Non-structural protein 8 (Protein ID: P0DTC8) of SARS-CoV-2 (Severe Acute Respiratory Syndrome-related coronavirus 2) for T cell assays (e.g. ELISPOT).
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SARS-COV-2 ORF8 1VIAL(25UG/PEPTIDE) 76511-260 RP30017 Each Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty