Would you like to log in again?
SARS-COV-2 SPIKE GLYCOPRO
Supplier: Genscript
Sars-Cov-2 Spike Glycopro, Th pool includes 316 peptides (delivered in two subpools of 158/158 peptides) derived from peptide scan (15mers with 11aa overlap) through entire Spike glycoprotein (P0DTC2) of SARS-CoV-2 (Severe Acute Respiratory Syndrome-related coronavirus 2) for T cell assays.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SARS-COV-2 SPIKE GLYCOPRO 76511-266 RP30020 Each Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty