Would you like to log in again?
SAA ANTIGEN T02002-10 10MG
Supplier: Genscript
Serum Amyloid A (SAA) is an acute-phase protein. Concentration of SAA in blood increases rapidly when tissue damage or inflammation occurs. It is considered as an early biomarker for diagnosis of many inflammatory diseases. SAA can also be used as an important indicator of infection, Size: 10mg
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SAA ANTIGEN T02002-10 10MG 76511-292 T02002-10 Each (10mg) Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty