Would you like to log in again?
H-FABP (HC29) 10MG
Supplier: Genscript
Heart-type fatty acid binding protein (H-FABP) encoded by FABP3 gene with a low molecular weight of 15 kDa. It released in large quantities into blood at an early stage of acute myocardial infarction (AMI). H-FABP serum marker with high sensitivity specificity for diagnosing AMI a, Size: 10mg
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
H-FABP (HC29) 10MG 76511-490 V00904-10 Each (10mg) Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty