Would you like to log in again?
TEV PROTEASE HIS >95% 10000IU
Supplier: Genscript
Tobacco Etch Virus Protease, Source: E. Coli, Purity: > 95% by SDS-PAGE analyses, Molecular Weight: 28.4kDa, observed by reducing SDS-PAGE.Storage: Recombinant Tobacco Etch Virus Protease remains stable up to 1year at -20C from date of receipt, Synonyms: rTEV, TEV, P1 protease, Size: 10000 IU
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TEV PROTEASE HIS >95% 10000IU 76512-418 Z03030-10K Each Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty