Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

NLS-Cas9-EGFP Nuclease (from E. coli)

Supplier: GenScript Biotech

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Z03393-50 Z03393-100
76513-840EA 196.46 USD InStock
76513-840 76513-838
NLS-Cas9-EGFP Nuclease (from E. coli)
Proteins and Peptides
GenCrispr NLS-Cas9-EGFP is a fusion protein, contains a nuclear localization signal (NLS) on its N terminal end, and an EGFP and a 6x(His) sequence on the C terminal end. Cas9 nuclease is an RNA-guided endonuclease that can catalyze cleavage of double stranded DNA. This kind of targeted nuclease is a powerful tool for genome editing with high precision.

  • DNA-free: no external DNA added to system
  • High cleavage efficiency: NLS ensures the efficient entry of the Cas9 protein into nuclei
  • Low off target effects: transient expression of Cas9 nuclease improves specificity of cleavage
  • Time-saving: no need for transcription and translation
  • Reduced labor: enrich cell populations for desired genome edits via EGFP-based FACS, the C-terminal His-tag increases the choice of detection methods for the fusion protein

Cas9 protein forms a highly stable ribonucleoprotein (RNP) complex with the guide RNA (gRNA) component of the CRISPR/Cas9 system. When Cas9 is expressed with an NLS sequence, the Cas9 RNP complex can localize to the nucleus immediately upon entering the cell. There is no requirement for in vivo transcription or translation, which improves the efficiency of this method over plasmid-based systems. Additionally, the Cas9 RNP complex is rapidly cleared from the cells minimizing the chance of off-target cleavage when compared to other systems.

Applications include screening for highly efficient and specific targeting gRNAs by in vitro DNA cleavage, in vivo gene editing when combined with a specific gRNA by electroporation or injection and enrichment of cell populations for desired genome edits via EGFP-based FACS.

Delivery information: GenCrispr NLS-Cas9-EGFP is supplied with 1X storage buffer (10 mM Tris, 300 mM NaCl, 0.1 mM EDTA, 1 mM DTT, 50% Glycerol pH7.4 at 25 °C).

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty