Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

MPU Series Connectors, CPC

Supplier: CPC (Colder Products Company)
MPU221639M MPU171639
76514-162EA 74.43 USD InStock
76514-162 76514-160
MPU Series Connectors, CPC
Fittings and Connectors
MPU series connector insert.

  • ³/₄ and 1" hose barb: Facilitates rapid fill and empty of bioprocessing bag
  • Locking feature: Guards against accidental disconnects
  • Sharp barb end: Minimizes fluid furbulence and dead space
  • Shrouded, leak-free seal and smooth, internal flow path
  • Protect valuable fluids and eliminate potential to contaminate fluid path

The MPU's twist-to-connect design features an easy-to-use locking mechanism that guards against accidental disconnects and provides a reliable, secure connection. A ³/₄ and 1" hose barb provides smooth, rapid media transfer.

Gamma Sterilization: Up to 50 kGy irradiation Autoclave: Up to 132 °C (270 °F), 60 minutes, up to 25 repetitions Sterilize uncoupled only.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty