Would you like to log in again?
GLOVES ESD GRAY/WHITE TOUCHSCREEN SIZE 6
Supplier: NEWELL BRANDS INC
Gloves, ESD, 18 guage, specifically developed to dissipate electrostatic charges for precise and repetitive tasks where dexterity, comfort and durability is required​. Touchscreen on thumb and index finger, Size: 6
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLOVES ESD GRAY/WHITE TOUCHSCREEN SIZE 6 76514-190 34524006 Case of 96 Retrieving
Pair Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty