Would you like to log in again?
J-Fill® Uno Dispensing System, Diversey
Supplier: Diversey
D5753036
76514-342CS 428.19 USD InStock
76514-342
J-Fill® Uno Dispensing System, Diversey
Soap Dispensers and Lotion Soap Dispensers
Accurately dispenses a single J-Fill product with the push of a button.
  • Rugged powder-coated steel construction resists corrosion and comes with lockable cabinet doors
  • Meets ASSE 1055-B back-flow prevention requirements
  • Design only allows J-Fill packaging to be used within dispenser
  • Uses super-concentrated and hyper-concentrated chemicals, reduces 'in-use' cost and solid waste disposal
  • Metering tip in each J-Fill cartridge ensures precise dilution ratios
Compact, lockable, wall-mounted single station dispensing unit. Accurately dispenses one J-Fill® product and rinse water into spray bottles or buckets/autoscrubbers with the push of a button. Rugged stainless steel construction.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty