Would you like to log in again?
Alpha-HP® Disinfectant Cleaner, Diversey
Supplier: Diversey
5549254
76514-344CS 131.01 USD InStock
76514-344
Alpha-HP® Disinfectant Cleaner, Diversey
Cleaning Agents Disinfectants
One product cleans and brightens surfaces safely and easily.
  • Bactericidal and virucidal, no-food contact sanitizer
  • Hydrogen peroxide active ingreadient breaks down to oxygen and water after use
  • Excellent cleaning performance and environmental profile
  • Meets daily cleaning and disinfection needs
  • For use on hard, inanimate, non-porous surfaces such as floors, walls, glazed procelain and plastic surfaces
An all-in-one, multi-surface disinfectant cleaner based on proprietary accelerated hydrogen peroxide (AHP®) technology. One product cleans and brightens surfaces safely and easily. Use on walls, countertops and most other water washable hard surfaces.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty