Would you like to log in again?
Oxivir® Five 16 Concentrate, Diversey
Supplier: Diversey
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
4963314 5019296
76514-350CS 133.9 USD InStock
76514-350 76514-338
Oxivir® Five 16 Concentrate, Diversey
Cleaning Agents Disinfectants
Delivers fast, effective cleaning performance.
  • Kills MRSA, VRE and Norovirus, and Canine Parvovirus
  • Based on proprietary hydrogen peroxide technology (AHP)
  • Disinfects in 5 minutes at 1:16 dilution
  • HMIS ratings are all zeros
  • For use on hard, inanimate, non-porous surfaces
  • Meets bloodborne pathogen standards for blood and body fluid clean up
A one-step disinfectant cleaner based on proprietary accelerated hydrogen peroxide (AHP®) technology to deliver fast, effective cleaning performance. At 1:16 dilution, disinfects in five minutes. Virucide, bactericide, fungicide, mildewcide and non-food contact sanitizer. Kills MRSA, Norovirus and Canine parvovirus. Concentrate delivers economical performance. Meets bloodborne pathogen standards for decontaminating blood and body fluids. Colorless with a characteristic scent.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty