Would you like to log in again?
Oxivir® Tb Wipes, Diversey
Supplier: Diversey
4599516 5388471
76514-356CS 364.24 USD InStock
76514-356 76514-358
Oxivir® Tb Wipes, Diversey
Wipes Disposable Wipes and Towels Disinfectant Wipes
Delivers fast, effective cleaning performance.
  • Virucide, bactericide, tuberculocide, fungicide and sanitizer
  • Effective against key pathogens
  • Kills HBV, HCV and HIV in 1 minute
  • Tuberculocidal in 5 minutes
  • Fungicidal in 3 minutes
  • No VOCs, no added fragrance, no NPEs
  • Environmentally responsible, active ingredient breaks down to oxygen and water
Ready-to-use disinfectant cleaner wipes based on proprietary hydrogen peroxide (AHP®) technology to deliver fast, effective cleaning performance. Disinfects in 60 seconds. Virucide, bactericide, tuberculocide, fungicide and non-food contact sanitizer. Kills MRSA and Norovirus. Meets bloodborne pathogen standards for decontaminating blood and body fluids. Colorless with a characteristic scent.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty