Would you like to log in again?
Reusable Altessa Grid Cleanroom Frocks with Zipper Closure, Uniform Technology
Supplier: Uniform Technology
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CFRZC-74LB-S CFRZC-74LB5XL CFRZC-74LB2XL CFRZC-74WH2XL CFRZC-74WH-XL CFRZC-74LB3XL CFRZC-74NV2XL CFRZC-74NV-S CFRZC-74WH-L CFRZC-74NV-XS CFRZC-74LB4XL CFRZC-74NV-L CFRZC-74LB-M CFRZC-74NV3XL CFRZC-74WH3XL CFRZC-74WH-M CFRZC-74WH5XL CFRZC-74LB-XL CFRZC-74NV4XL CFRZC-74WH-S CFRZC-74NV-M CFRZC-74WH4XL CFRZC-74LB-L CFRZC-74NV5XL CFRZC-74WH-XS CFRZC-74LB-XS CFRZC-74NV-XL
76515-684EA 38.95 USD InStock
76515-684 76515-718 76515-716 76515-732 76515-710 76515-698 76515-730 76515-696 76515-736 76515-714 76515-712 76515-734 76515-690 76515-694 76515-692 76515-728 76515-706 76515-704 76515-726 76515-708 76515-688 76515-720 76515-686 76515-702 76515-724 76515-722 76515-700
Reusable Altessa Grid Cleanroom Frocks with Zipper Closure, Uniform Technology
Workwear
Cleanroom frocks are manufactured from 99% polyester/1% carbon fabric.
  • Military collar
  • Raglan sleeves
  • ESD knit wrists
  • Coil zipper with top snaps
Suitable for ISO Class 5 or higher cleanrooms.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty