Would you like to log in again?
Reusable Altessa Grid Pull-Over Cleanroom Hoods, Uniform Technology
Supplier: Uniform Technology
CHPO-74WH-L CHPO-74WH-XL CHPO-74NV-XS CHPO-74NV-XL CHPO-74NV-M CHPO-74WH-XS CHPO-74WH-2XL CHPO-74NV-S CHPO-74WH-M CHPO-74NV-L CHPO-74WH-S CHPO-74NV-3XL CHPO-74NV-2XL CHPO-74WH-3XL
76268-138EA 42.03 USD InStock
76268-138 76515-794 76515-792 76268-142 76268-140 76515-806 76515-804 76515-808 76515-798 76515-796 76515-802 76515-812 76515-800 76515-810
Reusable Altessa Grid Pull-Over Cleanroom Hoods, Uniform Technology
Workwear
Cleanroom hoods are manufactured from 99% polyester/1% carbon fabric.
  • Pull-over, open face design
  • Vertical and horizontal snaps on back
  • Snaps at temples
  • Split skirt
Suitable for ISO Class 5 or higher cleanrooms.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty