Would you like to log in again?
Reusable Altessa Grid Snap-Front Cleanroom Hoods, Uniform Technology
Supplier: Uniform Technology
CHM-74WH-3XL CHM-74NV-M CHM-74WH-XS CHM-74NV-2XL CHM-74WH-M CHM-74WH-2XL CHM-74NV-XL CHM-74WH-L CHM-74NV-3XL CHM-74NV-S CHM-74NV-L CHM-74WH-S CHM-74WH-XL CHM-74NV-XS
76515-828EA 18.47 USD InStock
76515-828 76515-838 76515-816 76515-826 76515-814 76515-836 76515-818 76515-820 76515-830 76515-840 76515-824 76515-834 76515-822 76515-832
Reusable Altessa Grid Snap-Front Cleanroom Hoods, Uniform Technology
Workwear
Cleanroom hoods are manufactured from 99% polyester/1% carbon fabric.
  • Open face design
  • Snaps under chin
  • Vertical and horizontal snaps on back
  • Snaps at temples
Suitable for ISO Class 5 or higher cleanrooms.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty