Would you like to log in again?
Reusable Maxima ESD Cleanroom Boots with Hypalon Sole, Uniform Technology
Supplier: Uniform Technology
CB2-68WH-XL CB2-68WH-S CB2-68WH-L CB2-68WH-XS CB2-68WH-M CB2-68WH-2XL CB2-68WH-3XL
76515-904PR 35.76 USD InStock
76515-904 76515-914 76515-902 76515-908 76515-906 76515-912 76515-910
Reusable Maxima ESD Cleanroom Boots with Hypalon Sole, Uniform Technology
Workwear
Cleanroom boots are manufactured from 99% polyester/1% carbon fabric.
  • Color-coded by size webbing strap
  • Side release buckle
  • Hypalon sole
Suitable for ISO Class 4 or higher cleanrooms.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty