Would you like to log in again?
Reusable ESD StatStar Labcoats, Long, Snap Wrists, Uniform Technology
Supplier: Uniform Technology
BR51-44WH-2XL BR51-44RB-2XL BR51-44WH-3XL BR51-44RB-5XL BR51-44RB-4XL BR51-44WH-M BR51-44WH-4XL BR51-44RB-XL BR51-44RB-XS BR51-44WH-S BR51-44WH-L BR51-44RB-S BR51-44WH-5XL BR51-44WH-XS BR51-44RB-L BR51-44RB-3XL BR51-44WH-XL BR51-44RB-M
76516-068EA 41.62 USD InStock
76516-068 76516-094 76516-072 76516-082 76516-096 76516-074 76516-084 76516-076 76516-098 76516-086 76516-100 76516-078 76516-088 76516-066 76516-090 76516-092 76516-070 76516-080
Reusable ESD StatStar Labcoats, Long, Snap Wrists, Uniform Technology
Workwear
Reusable ESD labcoats are manufactured from 87% polyester/13% carbon.
  • Lapel collar
  • Snap front
  • 2 waist pockets, 1 chest pocket
  • Snap wrists
Reusable ANSI S20.20 ESD labcoat features grounding snaps on the waist pockets.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty