Would you like to log in again?
Reusable ESD StatMaster Labcoats, Long, Snap Wrists, Uniform Technology
Supplier: Uniform Technology
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
BR5-47NB-2XL BR5-47WH-M BR5-47WH-5XL BR5-47WH-L BR5-47WH-2XL BR5-47NB-M BR5-47WH-S BR5-47WH-3XL BR5-47NB-5XL BR5-47NB-XL BR5-47NB-XS BR5-47NB-L BR5-47NB-4XL BR5-47WH-XL BR5-47WH-XS BR5-47NB-3XL BR5-47NB-S BR5-47WH-4XL
76516-208EA 53.89 USD InStock
76516-208 76516-178 76516-200 76516-202 76516-204 76516-206 76516-182 76516-192 76516-184 76516-194 76516-186 76516-196 76516-174 76516-188 76516-198 76516-176 76516-180 76516-190
Reusable ESD StatMaster Labcoats, Long, Snap Wrists, Uniform Technology
Workwear
Reusable ESD labcoats are manufactured from 93% polyester/7% carbon.
  • Lapel collar
  • Snap front
  • 2 waist pockets, 1 chest pocket
  • Snap wrists
Reusable ANSI S20.20 ESD labcoat features grounding snaps on the waist pockets.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty