Would you like to log in again?
Reusable ESD StatMaster Labcoats, Short, ESD Knit Wrists, Uniform Technology
Supplier: Uniform Technology
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
BR4C-47NB-XL BR4C-47WH-2XL BR4C-47NB-4XL BR4C-47NB-S BR4C-47WH-5XL BR4C-47WH-XL BR4C-47WH-4XL BR4C-47WH-3XL BR4C-47NB-L BR4C-47NB-3XL BR4C-47NB-5XL BR4C-47WH-XS BR4C-47WH-L BR4C-47NB-2XL BR4C-47WH-M BR4C-47NB-XS BR4C-47WH-S BR4C-47NB-M
76516-212EA 41.48 USD InStock
76516-212 76516-234 76516-244 76516-222 76516-236 76516-214 76516-224 76516-238 76516-216 76516-226 76516-218 76516-228 76516-230 76516-240 76516-232 76516-210 76516-242 76516-220
Reusable ESD StatMaster Labcoats, Short, ESD Knit Wrists, Uniform Technology
Workwear
Reusable ESD labcoats are manufactured from 93% polyester/7% carbon.
  • Lapel collar
  • Snap front
  • 2 waist pockets, 1 chest pocket
  • ESD knit wrists
Reusable ANSI S20.20 ESD labcoat features grounding snaps on the waist pockets.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty