Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
PURELAB® Chorus 1 Complete Water Purification System, ELGA LabWater
Supplier: ELGA LabWater
PURELAB® Chorus 1
PC120COBPM1 SK-CSPPC120-B PC110COBPM1 SK-CSPPC120-A PC120COBPM1 PC110COBPM1 SK-CSPPC110-B SK-CSPPC110-A
76048-804EA 9687.69 USD InStock
76048-804 76516-418 76048-806 CA76048-804 76516-420 76516-414 CA76048-806 76516-416
PURELAB® Chorus 1 Complete Water Purification System, ELGA LabWater
Water Purification Systems Pure and Ultra Pure Water Systems
PURELAB Chorus 1 Complete provides a complete solution from tap to ultrapure water (Type I) direct from a potable water supply, and is ideal for laboratories needing up to 100 liters of 18.2 MΩ.cm ultrapure water. With its simple and ergonomic design and ease-of-use, water can be dispensed directly from the system or from a choice of additional Halo Dispensers.
  • Recirculation of purified water through our modular reservoir to maintain consistent peak water purity at 18.2 MΩ.cm
  • When fitted, PURELAB Chorus 1 Complete produces water which is free from biologically active impurities
  • Perfect single system solution for analytical and life science applications requiring 18.2 MΩ.cm
  • Front entry service doors provide quick and easy access to consumables
  • Designed to be modular and stackable to save space, whether wall-mounted or under the bench
  • Data Capture via USB for system performance validation and software updates
The modular nature of PURELAB Chorus 1 complete means that your dispense solutions can sit independently from the unit
Suggested applications include mass spectrometry, molecular biology, electrochemistry, atomic spectroscopy, liquid chromatography, gas chromatography, immunochemistry, general laboratory, spectrophotometry.
Certifications: UL listed. CE marked.
Accessories information: Additional accessories, and future consumables are sold separately.
Ordering information: Package includes the Chorus water system and standard installation Kit (LA672). A water storage reservoir is required for this system to operate. If required, additional kits can be purchased for alternative configurations, including wall mounting, stacking and combining of systems. It is highly recommended to contact your sales representative for additional information to meet your application, budget, and configuration needs.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
SERVICES
Services: ELGA LabWater provides several service options, such as Extended Warranty, Installation, Preventative Maintenance, and Validation. To learn more about these services, please search "ELGA Water Technical Service" in the website search field.
In addition,VWR CATALYST™ can provide an array of services, such as on-site IQ, OQ, PQ, Customized Protocol Generation, Equipment Calibration, Asset Management, and Certification Services (US only). Please call 888-793-2300 or email VWRCatalyst@vwr.com for further details.
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty