Would you like to log in again?
Multi-position Submersible Magnetic Stirrers, Crystal Industries
Supplier: Crystal
MS-06BSUA MS-15BSUA MS-04BSUA
76516-660EA 915.88 USD InStock
76516-660 76516-662 76516-664
Multi-position Submersible Magnetic Stirrers, Crystal Industries
Stirrers Magnetic Stirrers, Multi-position
These multi-position submersible magnetic stirrers feature a space saving design, ideal for stirring multiple size beakers or flasks simultaneously and have been constructed to operate safely in a water bath or CO₂ incubator.
  • Stir a variety of beaker or flask sizes simultaneously in a compact space
  • Waterproof body for submersible use in water baths
  • Five stirring levels provide excellent mixing capability
  • Sealed stainless steel body construction protects against intrusion when used in a CO₂ incubator
  • Two years replacement warranty
Multi-position magnetic stirrers are simple to use and save valuable bench space. Ideal for any laboratory that needs to stir a variety of beakers or flasks simultaneously. Constructed from high quality materials for safe, reliable operation in a water bath or CO₂ incubator.
Delivery information: Stir bars included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty