Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Multi-position Hotplate Stirrers, Crystal Industries
Supplier: Crystal
MS-05HA MS-15HA MS-10HA
76516-670EA 1156.9 USD InStock
76516-670 76516-672 76516-668
Multi-position Hotplate Stirrers, Crystal Industries
Stirrers Magnetic Hotplate Stirrers, Multi-position
These multi-position hotplate stirrers feature a space saving design, ideal for applications that require both stirring and heating.
  • Fast heating speed and excellent temperature uniformity
  • Heat and stir a variety of different sized beakers or flasks simultaneously
  • Exterior is aluminum alloy for high temperature resistance, corrosion protection and excellent insulation properties
  • Two-year replacement warranty
Multi-position hotplate stirrers are simple to use and save valuable bench space. Simply set the speed and temperature for the application. Ideal for any laboratory that needs heating and stirring in a variety different sized beakers or flasks. Constructed from high quality materials for reliable, quiet operation.
Delivery information: Stir bars included.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty