Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Laboratory Glassware Washer, PLW8505 Promo Bundle
Supplier: Miele
94003800
76518-114EA 10984.66 USD InStock
76518-114
Laboratory Glassware Washer, PLW8505 Promo Bundle
Glassware Washers
Promo bundle consisting of glassware washer PLW8505, APLW 097, APLW 096, ProCare 10AP and ProCare 30C with savings up to 15%.
  • 39 narrow-neck/lab glassware with injectors
  • Two integrated dispenser pumps with DI water connection
Includes steam condenser, stainless steel interior, glass door, liquid detergent and liquid acid neutralizer internal dispensing pumps, 115 V/60 Hz. Simple operation - LED color display with Touch on Glass Dispense process chemicals easily: Two integrated dispenser pumps With DI water connection - final rinse with demineralized water. Have everything in view thanks to the full-glass door.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty