Would you like to log in again?
CLEANROOM UPGRADE FOR 7400-33 MODERATE
Supplier: CARON PRODUCTS & SERVICE
Cleanroom Upgrade, For 7400-33 Moderate, series incubator to handle exterior wipedown with moderate cleaning chemicals, includes stainless steel exterior hinges, stainless steel hardware, and coated magnetic door trim
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CLEANROOM UPGRADE FOR 7400-33 MODERATE 76518-142 CLEN302 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty