Would you like to log in again?
CLEANROOM UPGRADE FOR 7400-25 AGGRESSIVE
Supplier: CARON PRODUCTS & SERVICE
Cleanroom Upgrade, For 7400-25 Aggressive, series incubator to handle exterior wipedown with aggressive cleaning chemicals, includes 316 stainless steel on all exterior surfaces as well as hinges and hardware, coated magnetic door trim, and capacitive touchscreen
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CLEANROOM UPGRADE FOR 7400-25 AGGRESSIVE 76518-144 CLEN303 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty