Would you like to log in again?
KIT WALL TRIM FOR 33CF
Supplier: CARON PRODUCTS & SERVICE
Kit, Wall Trim, For 33Cf, incubator recessed into a wall, Includes double vapor barrier frame seal and bottom coved skirt, Ideal for applications where the front of the incubator is installed in a cleanroom and the back is located outside the cleanroom
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
KIT WALL TRIM FOR 33CF 76518-158 WALL302 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty