Would you like to log in again?
WALL INSTALL PREP FOR CLEN303 7400-25/33
Supplier: CARON PRODUCTS & SERVICE
Wall Install Prep, For Clen303 7400-25/33, with CLEN303 stainless steel incubator (non-refrigerated), Includes 316 stainless steel split-cover access panel, power switch moved to the rear, and solid front bezel
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
WALL INSTALL PREP FOR CLEN303 7400-25/33 76518-164 WALL305 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty