Would you like to log in again?
VWR® Aloe-Coated Nitrile Gloves
Supplier: VWR
VWR®
14024421 14024422 14024423 14024419 14024420
76518-176CS 284.29 USD InStock
76518-176 76518-178 76518-172 76518-174 76518-180
VWR® Aloe-Coated Nitrile Gloves
Gloves
The innovative aloe-coated nitrile glove is designed to combine the durability of nitrile with the soothing properties of aloe vera. The aloe coating provides a moisturizing and nourishing experience, keeping hands hydrated and preventing dryness and irritation.
  • Aloe-coated
  • 3.5 mil finger thickness
  • Powder-free
  • 9.5" length
  • Food contact compliant
  • High comfort and flexibility
  • Textured fingers
Perfect for exceptional grip and tactile sensitivity, ensuring optimal performance in various industries such as healthcare, laboratory, food handling, and more.
Packaging: Gloves are packaged 200 per box, 10 boxes per case.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty