Would you like to log in again?
Aquasearcher™ AB41PH Bench Meter, Ohaus
Supplier: Ohaus
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
30589830 30589831 30589831
76518-248EA 1134.11 USD InStock
76518-248 470336-586 76518-250
Aquasearcher™ AB41PH Bench Meter, Ohaus
Electrochemical Meters pH and Multi-Parameter Meters
An advanced, research-grade benchtop pH meter offering accurate, repeatable results.
  • With selectable resolution from 0.1 to 0.001 and an intelligent i-Steward system, AB41PH is an excellent pH benchtop meter with high resolution and consistent accuracy
  • Equipped with a 6.5-inch large LCD display and a touch keypad, AB41PH offers a simple and precise control experience
  • The AB41PH features a 1000-item memory and password management for GLP documentation
  • Provides connectivity and data output capabilities with RS232 and USB interface
i-Steward Indicators include the pH electrode condition, 'Electrode Dirty/Broken', and reminder when meter needs recalibration. Selectable resolution from 0.1 to 0.001. Perform up to a 5-point pH calibration using automatic buffer recognition of 5 predefined buffer groups. Calibration log stores ten most recent calibrations. Compact stirrer can be used to uniformly mix solutions to get fast and repeatable results at no additional cost. With multi-function touch keypad, you can operate the meter as easy as using a smartphone.
All of the important information that you may need is shown on a 6.5” large display in multiple languages. Capture your critical readings with measurement stability indicator and selectable end-points modes – Auto-Stop holds a stable reading, Continuous shows changing readings and Timed logs data at specific intervals. A 1000-item memory for measurements and calibration trail makes for efficient data documentation. Password management is suitable for GLP documentation. Standard RS232 and USB interface allows for connection to external devices such as a printer or computer for transfer or storage of data.
Certifications: Product Safety: IEC/EN 61010-1. Electromagnetic Safety: IEC/EN 61326-1 Class B, basic environments; FCC Part 15 Class A; Canada ICES-003 Class A. Compliance Marks: CE, RCM. Others: WEEE, RoHS.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty