Would you like to log in again?
Aquasearcher™ AB33PH Bench Meter, Ohaus
Supplier: Ohaus
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Aquasearcher™
30589827 30589826 30589827
76518-258EA 766.53 USD InStock
76518-258 76518-256 470336-584
Aquasearcher™ AB33PH Bench Meter, Ohaus
Electrochemical Meters pH and Multi-Parameter Meters
Highly reliable and user-friendly pH benchtop meter for standard laboratory applications.
  • With multifunctional touch keypads, AB33PH makes measurement simple and fast within 3 steps
  • The intelligent i-Steward monitors the condition of electrodes, ensuring the accuracy of the results
  • Auto endpoint mode and auto buffer recognition makes calibration easy
  • A 1000-item memory for pH measurements and calibration trail makes for efficient data documentation
  • A user-friendly bench meter with an innovative design, including 6.5 in. large LCD display & adjustable stand-alone electrode holder
  • Multiple connectivity capabilities include RS232 and USB interface
i-Steward Indicators include the pH electrode condition, 'Electrode Dirty/Broken', and reminder when meter needs recalibration. On-screen text prompts, menu-specific function keys and a multilanguage interface shown on a 6.5” large display. Three capacitive touch keys that change based upon use ensures the buttons are never clogged with sample residue. Most of the operation can be done within 3 steps. Auto-Stop holds a stable reading. Continuous shows changing readings, perform up to a 3-point pH calibration using automatic buffer recognition of 3 predefined buffer groups, selectable cell constant allows for use with 2- or 4-cell conductivity cells to measure from ultrapure water to sewage.
Adjustable TDS Factor - The factor that relates conductivity to total dissolved solids is based on the type of sample being measured. To have correct temperature compensation when measuring conductivity, the linear compensation coefficient adjusts by automatic temperature compensation. Selectable reference temperatures of 20 or 25 °C for temperature compensated conductivity readings. A 1000-item memory for measurements and calibration trail makes for efficient data documentation. Records are associated with date and time for good traceability. Standard RS232 and USB interface allows for connection to external devices such as a printer or computer for transfer and storage of data. A standalone adjustable electrode holder provides full flexibility for different types of electrodes
Certifications: • Product Safety: IEC/EN 61010-1 • Electromagnetic Safety: IEC/EN 61326-1 Class B, basic environments; FCC Part 15 Class A; Canada ICES-003 Class A • Compliance Marks: CE, RCM • Others: WEEE, RoHS
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty