Would you like to log in again?
Brady® SPC® Absorbents Oil-Only 14 gal. Spill Kit Refill, Brady
Supplier: Brady
SKO14-R
76518-274KT 120.74 USD InStock
76518-274
Brady® SPC® Absorbents Oil-Only 14 gal. Spill Kit Refill, Brady
Spill Control Materials
SPC® Spill Kits from Brady® combine absorbents, accessories, PPE and instructional guidelines so you're prepared when a spill occurs.
  • Absorption capacity 13 gal.
  • Oil Only spill control
Refill kit only (container SKO-14 sold separately). White absorbents pick up petroleum-based fluids, paints and non-water-soluble chemicals while repelling water. Great for indoor and outdoor use. Improve worker productivity and efficiency by placing spill kits around your facility close to areas prone to spills. Spill kits are ideal for fast response to critical accidents.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty