Would you like to log in again?
Brady® SPC® Absorbents Chemical 14 gal. Spill Kit Refill, Brady
Supplier: Brady
SKH14-R
76518-276KT 123.58 USD InStock
76518-276
Brady® SPC® Absorbents Chemical 14 gal. Spill Kit Refill, Brady
Spill Control Materials
SPC® Spill Kits from Brady® combine absorbents, accessories, PPE and instructional guidelines so yourre prepared when a spill occurs.
  • Absorption capacity 10.1 gal.
  • Chemical spill control
Refill kit only (container SKH-14 sold separately). Surfactant-treated, brightly colored absorbent picks up a wide range of aggressive chemicals while remaining chemically inert. Improve worker productivity and efficiency by placing spill kits around your facility close to areas prone to spills. Spill kits are ideal for fast response to critical accidents.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty