Would you like to log in again?
Brady® SPC® Absorbents Trade Agreements Act (TAA) Compliant 14 gal. Drum Absorbent Kits, Brady
Supplier: Brady
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SKA14-TAA
76518-280KT 211.18 USD InStock
76518-280
Brady® SPC® Absorbents Trade Agreements Act (TAA) Compliant 14 gal. Drum Absorbent Kits, Brady
Spill Control Materials
SPC® Spill Kits from Brady® combine absorbents, accessories, PPE and instructions in a space-saving container so you're prepared when a spill occurs.
  • Absorption capacity 12 gal.
  • TAA Compliant
  • Universal spill control
Highly versatile, gray absorbents pick up water-based, petroleum-based and non-aggressive chemical fluids. Yellow PVC bag offers high-visibility storage so spill-control items can be quickly located in an emergency situation. TAA compliant: manufactured or substantially transformed in a designated country, appropriate for GSA Schedule Contracts. Improve worker productivity and efficiency by placing spill kits around your facility close to areas prone to spills.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty