Would you like to log in again?
VWR® B2-Series Balances with Calibration Certificate
Supplier: VWR
VWR®
VWR-403B2/C VWR-203B2/C VWR-210B2/C VWR-503B2/C VWR-120B2/C VWR-60B2/C VWR-5002B2/C VWR-4002B2/C
76519-310EA 1731.72 USD InStock
76519-310 76378-408 76519-314 76519-312 76378-446 76378-444 76378-412 76378-410
VWR® B2-Series Balances with Calibration Certificate
Balances and Scales
The VWR® B2-Series/C balances which include an ISO/IEC 17025:2017 Calibration Certificate are the answer for an organizations regulatory and compliance needs.
  • ISO/IEC 17025:2017 Accredited Calibration and Testing Laboratory issues Calibration Certificate
  • Complete calibration data reported
  • Reliable results
  • RS232 interface
  • External calibration
  • Multiple weighing modes
  • Five year warranty
The balances include a Certificate of Calibration showing the full testing data from an ISO/IEC 17025:2017 accredited Calibration and Testing Laboratory utilizing NIST-traceable weights.
The B2-Series balances incorporate state-of-the-art features for advanced weighing applications. These economical balances offer a variety of standard features, such as external calibration, menu-driven operation, multiple weighing modes, and various application modes.
The 0.1 and 1 mg readability models feature a fixed draft shield (which can't be removed).
Certifications: ISO/IEC 17025:2017 Accredited Calibration and Testing Laboratory issues Calibration Certificate; CE marked.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty