Would you like to log in again?
GLOVES HYFLEX 11561VP SIZE 8.0 PK6
Supplier: Ansell Healthcare
Gloves, industrial, HyFlex 11-561VP, with 100% greater durability, thinnest, lightest ISO C/ANSI A3-level, Grey, Nitrile, Knitted, Knitwrist, Palm coated, 15gauge, 210-285mm, Darkblue, HPPE/Spandex/Polyester/Nylon/HPPE, Silicone free, Size: 8
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLOVES HYFLEX 11561VP SIZE 8.0 PK6 76519-338 11561VP080 Case of 864 Retrieving Restricted
Pack of 6 Retrieving Restricted
Restricted To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty