Would you like to log in again?
RINGERS R167 SIZE 11.0 CS1
Supplier: Ansell Healthcare
Ringers, R167 Cut & Sew, HIViz, Impact Protection, cut protection, synthetic red leather padded palm, touchscreen compatibility, Flexible knuckle TPR design, Rubber pull tab, Reinforced thmb, index & middlefingertips, Size: 11
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
RINGERS R167 SIZE 11.0 CS1 76519-428 167-11 Case of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty