Would you like to log in again?
FLATMOP POLYSORB NO FOAM 9IN WIDTH CS50
Supplier: MICRONOVA MFG INC
PolySorb FlatMop, No Foam, 9in width, 100% Textured Polyester, no interior foam. Texture assists in removing residues and dispersing disinfectant. Irradiated.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FLATMOP POLYSORB NO FOAM 9IN WIDTH CS50 76519-526 MZSM11-9NS Case of 50 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty