Self-sealing Tyvek® 1422A autoclave bags with steam indicators can be used as either the inner, or outer bag for the steam sterilization of pharmaceutical change parts.
- Manufactured and packaged in a controlled cleanroom environment
- Convenient self-sealing closure system
- Tear and puncture resistant, hydrophobic and lint-free
- Superior to paper based autoclave pouches and wraps
- Fast and consistent for equipment preparation area
- Strong factory seals
- Exceptional microbial barrier
Standardize and simplify the way equipment is wrapped for steam sterilization. The use of these self-sealing bags will also save valuable time in the equipment prep area.
The drapeable nature of the 1422A Tyvek® material is ideal for wrapping equipment parts where one may need to 'feel through' the bag; especially when lifting, or handling the wrapped part.
Since 100% of the surface area allows for steam penetration and drying, each load can be configured for maximum efficieny and throughput.
Certifications: Certificates of Conformance sent with each shipment.