Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tyvek® 1422A Autoclave Bags with Steam Indicator, Self-Seal, Keystone Cleanroom Products

Supplier: Keystone Cleanroom Products

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

BGT2A3060SS BGT2A1232SS BGT2A1820SS BGT2A1230SS BGT2A2030SS BGT2A0815SS BGT2A2044SS BGT2A1418SS BGT2A0713SS BGT2A4024SS BGT2A1612SS BGT2A2032SS BGT2A1526SS BGT2A1216SS BGT2A1543SS BGT2A3664SS BGT2A3838SS BGT2A4848SS BGT2A1212SS BGT2A0612SS BGT2A1062SS BGT2A3232SS BGT2A1648SS BGT2A4072SS BGT2A4242SS BGT2A1638SS BGT2A1448SS BGT2A0404SS BGT2A1236SS BGT2A1018SS BGT2A1246SS BGT2A1632SS BGT2A5054SS BGT2A4040SS BGT2A1630SS BGT2A1424SS BGT2A2440SS BGT2A4416SS BGT2A0638SS BGT2A0610SS BGT2A2048SS BGT2A3646SS BGT2A1024SS BGT2A3648SS BGT2A3248SS BGT2A1420SS BGT2A4864SS BGT2A1030SS BGT2A2424SS BGT2A1644SS BGT2A2036SS BGT2A1870SS BGT2A1836SS BGT2A1830SS BGT2A3624SS BGT2A1436SS BGT2A3672SS BGT2A1840SS BGT2A1228SS BGT2A1848SS BGT2A0205SS BGT2A2224SS BGT2A2634SS BGT2A1214SS BGT2A1616SS BGT2A3636SS BGT2A1218SS BGT2A0428SS BGT2A1020SS BGT2A2436SS BGT2A1832SS BGT2A0416SS BGT2A3028SS BGT2A1636SS BGT2A3660SS BGT2A1414SS BGT2A1824SS BGT2A0707SS BGT2A1604SS BGT2A3456SS BGT2A1234SS BGT2A2646SS BGT2A3030SS BGT2A0709SS BGT2A2024SS BGT2A0811SS BGT2A2452SS BGT2A1056SS BGT2A0409SS BGT2A1022SS BGT2A2832SS BGT2A3642SS BGT2A6250SS BGT2A1222SS BGT2A0615SS BGT2A09-512SS BGT2A2462SS BGT2A2828SS BGT2A2430SS BGT2A1240SS BGT2A2834SS BGT2A1248SS BGT2A0608SS BGT2A1048SS BGT2A2628SS BGT2A3628SS BGT2A1416SS BGT2A1220SS BGT2A1224SS BGT2A1438SS BGT2A0616SS BGT2A1854SS BGT2A1242SS BGT2A2844SS BGT2A1620SS BGT2A4850SS BGT2A2020SS BGT2A2040SS BGT2A0820SS BGT2A0426SS BGT2A3016SS BGT2A4048SS BGT2A4244SS BGT2A1015SS BGT2A3412SS BGT2A2448SS BGT2A4060SS BGT2A1624SS BGT2A3434SS BGT2A0430SS BGT2A3020SS BGT2A4450SS BGT2A2046SS BGT2A1440SS BGT2A1426SS
76519-750CS 3377.54 USD InStock
76519-750 76519-596 76519-594 76519-754 76519-632 76519-752 76519-598 76519-630 76519-758 76519-636 76519-756 76519-634 76519-638 76519-592 76519-590 76519-584 76519-582 76519-622 76519-588 76519-742 76519-620 76519-586 76519-740 76519-626 76519-746 76519-624 76519-744 76519-628 76519-748 76519-818 76519-772 76519-650 76519-770 76519-776 76519-654 76519-774 76519-652 76519-812 76519-658 76519-778 76519-810 76519-656 76519-816 76519-814 76519-808 76519-640 76519-760 76519-644 76519-764 76519-642 76519-762 76519-802 76519-648 76519-800 76519-646 76519-766 76519-806 76519-804 76519-680 76519-794 76519-672 76519-792 76519-670 76519-710 76519-798 76519-676 76519-796 76519-674 76519-714 76519-558 76519-712 76519-678 76519-718 76519-716 76519-708 76519-790 76519-662 76519-782 76519-660 76519-780 76519-820 76519-666 76519-786 76519-664 76519-784 76519-824 76519-702 76519-668 76519-822 76519-700 76519-788 76519-828 76519-706 76519-826 76519-704 76519-580 76519-574 76519-694 76519-572 76519-692 76519-732 76519-578 76519-698 76519-610 76519-576 76519-730 76519-696 76519-736 76519-614 76519-734 76519-612 76519-618 76519-738 76519-616 76519-570 76519-690 76519-684 76519-562 76519-682 76519-560 76519-688 76519-600 76519-720 76519-566 76519-686 76519-564 76519-604 76519-724 76519-602 76519-568 76519-722 76519-608 76519-728 76519-606 76519-726
Tyvek® 1422A Autoclave Bags with Steam Indicator, Self-Seal, Keystone Cleanroom Products
Bags Sterilization Pouches
Self-sealing Tyvek® 1422A autoclave bags with steam indicators can be used as either the inner, or outer bag for the steam sterilization of pharmaceutical change parts.

  • Manufactured and packaged in a controlled cleanroom environment
  • Convenient self-sealing closure system
  • Tear and puncture resistant, hydrophobic and lint-free
  • Superior to paper based autoclave pouches and wraps
  • Fast and consistent for equipment preparation area
  • Strong factory seals
  • Exceptional microbial barrier

Standardize and simplify the way equipment is wrapped for steam sterilization. The use of these self-sealing bags will also save valuable time in the equipment prep area.

The drapeable nature of the 1422A Tyvek® material is ideal for wrapping equipment parts where one may need to 'feel through' the bag; especially when lifting, or handling the wrapped part.

Since 100% of the surface area allows for steam penetration and drying, each load can be configured for maximum efficieny and throughput.

Certifications: Certificates of Conformance sent with each shipment.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty