Would you like to log in again?
Autoclave Wrappers for Steam, EtO or Gamma Sterilization, Blue BHD, Keystone Cleanroom Products
Supplier: Keystone Cleanroom Products
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
BHDSHEET0303 BHDSHEET0202 BHDSHEET01.5 BHDSHEET0606 BHDSHEET0404
76519-830CS 250 USD InStock
76519-830 76519-834 76519-832 76519-838 76519-836
Autoclave Wrappers for Steam, EtO or Gamma Sterilization, Blue BHD, Keystone Cleanroom Products
Bags Sterilization Pouches
When secured with autoclave tape or elasticized bands, Keystone Cleanroom Product’s Blue BHD wrappers provide an effective microbial barrier from external sources of contamination.
  • Designed for steam sterilization, but also compatible with EtO or gamma irradiation
  • Treated to impart water and alcohol repellency
  • Wet formed, lightweight non-woven fabric comprised of natural wood pulp and polyester fibers bonded with a resin binder
  • Superior bacterial barrier properties compared to linen
  • Sterilization tapes adhere firmly and remove easily
  • Ideal choice for for light trays, small basins and towels
  • Resists tearing and punctures
  • Does not contain any natural latex rubber
These versatile wraps offer fluid repellency and provide an excellent barrier against airborne bacteria. The soft, drapable non-woven blend of cellulose and polyester is virtually lint-free and helps protect wrapped contents from contamination.
Certifications: Certificates of Conformance with each shipment.
Packaging: Processed and packaged in ISO certified cleanrooms.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty