Would you like to log in again?
VWR® Wi-Fi Digital Data Logger (DDL) for Ultra Low Temperature Freezers (Dual Probe)
Supplier: Avantor
VWR®
CP-WF-DDL-18(-80)
76520-148EA 783.4 USD InStock
76520-148
VWR® Wi-Fi Digital Data Logger (DDL) for Ultra Low Temperature Freezers (Dual Probe)
Data Loggers
This Wi-Fi data logger is compliant with CDC guidelines. It includes a buffered probe in product simulated solution, a detachable dual probe (can be used with only one), audible/visual alarms, and temperature data via Wi-Fi network. Device holds 3 years of data storage and comes with a 3 years certificate of calibration.
  • Material: Assorted materials
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty